Learn More
ACROBiosystems Biotinylated Human HLA-C*08:01&B2M Monomer Protein (Peptide free, MALS verified)
Catalog No. 502719434
Encompass_Preferred
Biotinylated Human HLA-C*08:01&B2M Monomer Protein (HLM-H82Es) is expressed from human 293 cells (HEK293). It contains AA Cys 25 - Ile 308 (HLA-C*08:01) & Ile 21 - Met 119 (B2M) (Accession # QOC77848.1 (HLA-C*08:01) & P61769 (B2M)).
Catalog No. 50-271-9434 Supplier ACROBiosystems Supplier No. HLMH82ES25UG
Biotinylated Human HLA-C*08:01&B2M Monomer Protein (HLM-H82Es) is expressed from human 293 cells (HEK293). It contains AA Cys 25 - Ile 308 (HLA-C*08:01) & Ile 21 - Met 119 (B2M) (Accession # QOC77848.1 (HLA-C*08:01) & P61769 (B2M)).