Learn More
Sino Biological Anti-HLA-DR Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 68038XM01F100
Encompass_Preferred
The monoclonal antibody was purified by affinity chromatography. Then it was conjugated with FITC under optimum conditions, and the unreacted FITC was removed.
Catalog No. 50-241-2559