Learn More
Sino Biological Anti-HLA-DR Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 68038XM01P100
Encompass_Preferred
The monoclonal antibody was purified by affinity chromatography. Then it was conjugated with PE under optimum conditions, and the unreacted PE was removed.
Catalog No. 50-241-1962