Learn More
BIOHIPPO Human HLA-DR Microsample
SDP
Catalog No. 5000315692
Encompass
Human HLA-DR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HLA-DR in serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HLA-DR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HLA-DR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HLA-DR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15692 Supplier BIOHIPPO Supplier No. BHE15210652
Human HLA-DR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HLA-DR in serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HLA-DR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HLA-DR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HLA-DR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color