Learn More
Sino Biological Anti-HMGB1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100930T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human HMGB1, and purified by antigen affinity chromatography.
Catalog No. 50-255-5482