Learn More
Sino Biological Recombinant Human HMGB1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10326H08H1100
Encompass_Preferred
A DNA sequence encoding the Human HMGB1 (NP_002119.1) (Gly2-Glu215) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5380