Learn More
Sino Biological Recombinant Human HMGB1 Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 10326H49BB100
Encompass_Preferred
A DNA sequence encoding the Human HMGB1 (NP_002119.1) (Met1-Glu215) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5379