Learn More
BIOHIPPO Human HNE Microsample
SDP
Catalog No. 5000314805
Encompass
Human HNE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HNE in Serum plasma tissue homogenates It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 22 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HNE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HNE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HNE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14805 Supplier BIOHIPPO Supplier No. BHE15212427
Human HNE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HNE in Serum plasma tissue homogenates It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 22 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HNE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HNE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HNE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color