Learn More
BIOHIPPO Human HNF1b Microsample
SDP
Catalog No. 5000315016
Encompass
Human HNF1b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HNF1b in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 05 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HNF1b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HNF1b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HNF1b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15016 Supplier BIOHIPPO Supplier No. BHE15211476
Human HNF1b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HNF1b in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 05 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HNF1b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HNF1b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HNF1b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color