Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HO-1 ELISA Kit

Catalog No. EEL060
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL060 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL060 Supplier Invitrogen™ Supplier No. EEL060
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Heme Oxygenase 1 (HO1) ELISA quantitates HO1 in serum, plasma, and other biological fluids The Human Heme Oxygenase 1 (HO1) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09601
Assay Range 0.31-20 ng/mL
Assay Sensitivity 0.19 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Heme oxygenase
Sample Type Plasma, Serum
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 proteinbK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein;
Gene ID (Entrez) 3162
Gene Symbol HMOX1
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target HO-1
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.