Learn More
Bioworld HUMAN HOLO TRANSFERRIN
Supplier:  Bioworld 220603551
Encompass_Preferred
Human Holo Transferrin Source: Human Background: Transferrin is the iron-transport protein of vertebrate serum and donates iron to cells through interaction with a specific membrane receptor, CD71. Transferrin appears to be indispens...
Catalog No. 50-253-452