Learn More
Biomatik Corporation Human Homeodomain Interacting Protein Kinase 2 (HIPK2) ELISA Kit
Supplier:  Biomatik Corporation EKU08403
Encompass_Preferred
Human Homeodomain Interacting Protein Kinase 2 (HIPK2) ELISA Kit for detecting HIPK2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H2X6. Gene ID: 28996. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8670