Learn More
Sino Biological Anti-15-PGDH Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11205MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HPGD (rh HPGD; Catalog#11205-H08E; NP_000851.2; Met 1-Gln 266). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2116