Learn More
Sino Biological Recombinant Anti-Hemopexin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10870R036100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Hemopexin / HPX (rh Hemopexin / HPX; Catalog#10870-H08H; NP_000604.1; Met 1-His 462).
Catalog No. 50-241-0557