Learn More
Sino Biological Anti-Hemopexin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10870MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HPX / Hemopexin (rhHPX; Catalog#10870-H08H; Met 1-His 462; NP_000604.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9664