Learn More
Sino Biological Anti-Hemopexin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10870MM0820
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Hemopexin (rh Hemopexin; Catalog#10870-H08H; NP_000604.1; Met1-His462). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7872