Learn More
Sino Biological Anti-Hemopexin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10870RP03100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HPX / Hemopexin (rh HPX; Catalog#10870-H08H; Met 1-His 462; NP_000604.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8506