Learn More
BIORTUS USA INC Human HRAS Protein, biotinylated/GMPPNP_loaded, 100UG
Catalog No. NC2953323
Encompass_Preferred
Gene Name: HRAS. Uniprot: P01112. Construct: biotinylated-Avi-Hras(M1-L188),GMPPNP loaded. Expression Host: E. coli. Purity: 90%.
Catalog No. NC2953323 Supplier BIORTUS USA INC Supplier No. BP310820MB
Gene Name: HRAS. Uniprot: P01112. Construct: biotinylated-Avi-Hras(M1-L188),GMPPNP loaded. Expression Host: E. coli. Purity: 90%.