Learn More
Sino Biological Human HRAS PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12059RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HRAS (rh HRAS; Catalog#12059-H08B; P01112; Met1-Cys186). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3666