Learn More
Sino Biological Anti-HRG/HPRG Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10836MM0120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HPRG (rhHPRG; Catalog#10836-H08H; Met 1-Lys 525; NP_000403.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0419