Learn More
Sino Biological Anti-HRG/HPRG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200602T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human HRG / HPRG fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1746