Learn More
Sino Biological Anti-HRG/HPRG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10836T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HRG/HPRG (rh HRG/HPRG; Catalog#10836-H08H; NP_000403.1; Met1-Lys525). HRG/HPRG specific IgG was purified by Human HRG/HPRG affinity chromatography.
Catalog No. 50-255-5140