Learn More
BIOHIPPO Human hs-cTnI Microsample
SDP
Catalog No. 5000315740
Encompass
Human hs-cTnI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of hs-cTnI in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 9 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human hs-cTnI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human hs-cTnI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human hs-cTnI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15740 Supplier BIOHIPPO Supplier No. BHE15210591
Human hs-cTnI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of hs-cTnI in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 9 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human hs-cTnI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human hs-cTnI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human hs-cTnI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color