Learn More
BIOHIPPO Human hs Microsample
SDP
Catalog No. 5000315689
Encompass
Human hs Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of hs in Serum plasma and other biological fluids It provides a detection range of 4 69-300 pg/mL with a sensitivity of 1 42 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human hs Ab1-40 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human hs Ab1-40 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human hs Ab1-40 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15689 Supplier BIOHIPPO Supplier No. BHE15210607
Human hs Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of hs in Serum plasma and other biological fluids It provides a detection range of 4 69-300 pg/mL with a sensitivity of 1 42 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human hs Ab1-40 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human hs Ab1-40 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human hs Ab1-40 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color