Learn More
Biomatik Corporation Human HS (Heparan Sulfate) ELISA Kit
Supplier:  Biomatik Corporation EKF58484
Encompass_Preferred
Human HS (Heparan Sulfate) ELISA Kit for detecting HS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000ng/ml. Sensitivity: 46.9ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Heparan Sulfate. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9177