Learn More
Raybiotech Inc C-Series Human Heat Shock Protein Antibody Array Kit.
Supplier:  Raybiotech Inc AAHHSP14
Encompass_Preferred
Detects 9 Human heat shock proteins. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: HSP10, HSP27, HSP32, HSP40, HSP60, HSP70, HSP90, GRP75, Ubiquitin+1
Catalog No. 50-194-8589