Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human HSP90 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP276643
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276643 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276643 Supplier Novus Biologicals™ Supplier No. NBP276643

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Human HSP90 ELISA Kit (Chemiluminescence) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human HSP90 ELISA Kit (Chemiluminescence) measures human HSP90 in serum, plasma and other biological fluids.

Specifications

Assay Range 62.5 to 4000 pg/mL
Assay Sensitivity 37.5 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Biological Fluids, Plasma, Serum
Specificity This kit recognizes Human HSP-90 in samples. No significant cross-reactivity or interference between Human HSP-90 and analogues was observed.
Gene ID (Entrez) 3320
Gene Symbol HSP90AA1
Host Species Human
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.