Learn More
Sino Biological Recombinant Human HSP90-alpha / HSP90AA1 Protein, HPLC-verified 1
Supplier:  Sino Biological 11445HNCE1
Encompass_Preferred
A DNA sequence encoding the human HSP90 isoform 2 (NP_005339.3) C-terminal segment, corresponding to amino acid sequence (Glu 535-Asp 732) was expressed and purified, with two additional aa (Gly & Pro) at the N terminus.
Catalog No. 50-224-6940