Learn More
Sino Biological Anti-GRP94/HSP90B1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100613T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human GRP94/HSP90B1, and purified by antigen affinity chromatography.
Catalog No. 50-241-2366