Learn More
Sino Biological Anti-HSP70 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11660MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HSP70 (rh HSP70; Catalog#11660-H07B; NP_005337.2; Ala2-Asp641). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4643