Learn More
Sino Biological Human HSPA5 PAb 100UL 102056-T46-100
Supplier:  Sino Biological 102056T46100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GRP78/HSPA5, and purified by antigen affinity chromatography.
Catalog No. 50-240-8697