Learn More
Sino Biological Anti-Hsp27/HSPB1 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10351MM01H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human HSP27 (rh HSP27; Catalog#10351-H08E; NP_001532.1; Met 1 – Lys 205). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1599