Learn More
Sino Biological Human HSPB2 ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502373396
Encompass_Preferred
Each vial contains 13 ng of recombinant HSP27. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.6 ng/mL is recommended.
Catalog No. 50-237-3396 Supplier Sino Biological Supplier No. SEK103515
Each vial contains 13 ng of recombinant HSP27. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.6 ng/mL is recommended.