Learn More
Biomatik Corporation Human HSPD1/HSP-60 (60 kDa heat shock protein, mitochondrial) ELISA Kit
Supplier:  Biomatik Corporation EKF58769
Encompass_Preferred
Human HSPD1/HSP-60 (60 kDa heat shock protein, mitochondrial) ELISA Kit for detecting HSPD1; HSP-60 in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10809. Target Synonyms: HSP-60; HSPD1; Heat Shock Protein 60; cpn60; GROEL; SPG13; 60 kDa chaperonin; Chaperonin 60; heat shock 60kD protein 1; chaperonin; Heat shock protein 60; heat shock protein 65; HLD4; Hsp60; HSP65; HuCHA60; P60 lymphocyte protein; short heat shock protein 60 Hsp60s1; spastic paraplegia 13; autosomal dominant. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9162