Learn More
Sino Biological Anti-HtrA2/Omi Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10619T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HtrA2 (rh HtrA2; Catalog#10619-H08E; O43464-1; Ala134-Glu458). HtrA2 specific IgG was purified by Human HtrA2 affinity chromatography.
Catalog No. 50-241-0641