Learn More
Sino Biological Human HTRA2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10619T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HtrA2 / Omi (rh HtrA2 / Omi; Catalog#10619-H08E; O43464-1; Ala134-Glu458). HtrA2 / Omi specific IgG was purified by Human HtrA2 / Omi affinity chromatography.
Catalog No. 50-237-3768