Learn More
Biomatik Corporation Human Huntingtin Interacting Protein 1 (HIP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11774
Encompass_Preferred
Human Huntingtin Interacting Protein 1 (HIP1) ELISA Kit for detecting HIP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.129ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00291. Gene ID: 3092. Target Synonyms: ILWEQ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7502