Learn More
Biomatik Corporation Human Huntingtin Interacting Protein 1 Related Protein (HIP1R) ELISA Kit
Supplier:  Biomatik Corporation EKU10989
Encompass_Preferred
Human Huntingtin Interacting Protein 1 Related Protein (HIP1R) ELISA Kit for detecting HIP1R in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75146. Gene ID: 9026. Target Synonyms: HIP12; HIP3; ILWEQ; Huntingtin Interacting Protein 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7717