Learn More
American Research Products Inc Human Hyaluronidase PH 20 SPA
Catalog No. 5000328245
Encompass
Human Hyaluronidase PH-20 (SPAM1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 39 pg/mL Sample cell culture supernates Method Sandwich
Catalog No. 50-003-28245 Supplier American Research Products Inc Supplier No. CSBEL022471HUIS
Human Hyaluronidase PH-20 (SPAM1) ELISA kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 39 pg/mL Sample cell culture supernates Method Sandwich