Learn More
Biomatik Corporation Human Hydroxymethylbilane Synthase (HMBS) CLIA Kit
Supplier:  Biomatik Corporation EKU12039
Encompass_Preferred
Human Hydroxymethylbilane Synthase (HMBS) CLIA Kit for detecting HMBS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 68.6-50,000pg/ml. Sensitivity: 25.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P08397. Gene ID: 3145. Target Synonyms: PBG-D; PBGD; UPS; Hydroxymethylbilane Synthase; Uroporphyrinogen I Synthase; Porphobilinogen deaminase; Pre-uroporphyrinogen synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5438