Learn More
American Research Products Inc Human HydroXymethylglutaryl Co
Catalog No. 5000328594
Encompass
Human HydroXymethylglutaryl CoA Reductase (HMG-CoAR) ELISA kit from CUSABIO Detection Range 0 78 ng/mL- 50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28594 Supplier American Research Products Inc Supplier No. CSBE15964H
Human HydroXymethylglutaryl CoA Reductase (HMG-CoAR) ELISA kit from CUSABIO Detection Range 0 78 ng/mL- 50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich