Learn More
Biomatik Corporation Human Hydroxymethylglutaryl Coenzyme A Synthase (HMGCS) ELISA Kit
Supplier:  Biomatik Corporation EKU04856
Encompass_Preferred
Human Hydroxymethylglutaryl Coenzyme A Synthase (HMGCS) ELISA Kit for detecting HMGCS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01581. Gene ID: 3157. Target Synonyms: HMGCS1; 3-Hydroxy-3-Methylglutaryl-Coenzyme A Synthase 1,Soluble; HMG-CoA Synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6982