Learn More
Biomatik Corporation Human Hypoxia Inducible Factor 1 Alpha (HIF1a) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11926
Encompass_Preferred
Human Hypoxia Inducible Factor 1 Alpha (HIF1a) Mini sample ELISA Kit for detecting HIF1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16665. Gene ID: 3091. Target Synonyms: HIF1-A; MOP1; PASD8; Basic Helix-Loop-Helix Transcription Factor; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; Class E basic helix-loop-helix protein 78. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6948