Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human I-309 (CCL1) Recombinant Protein, PeproTech®

Catalog No. 300372UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-37-2UG 2 μg
30037100UG 100 μg
300-37-10UG 10 μg
30037250UG 250 μg
30037500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 300-37-2UG Supplier Gibco™ Supplier No. 300372UG
  • $92.50
    $69.65 / Each of 1

    Save $22.85

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human I-309 is an 8.5 kDa protein consisting of 74 amino acid residues that include six cysteine residues, of which four are the highly conserved cysteine residues present in CC chemokines. Unlike some CC chemokines, whose biologically active form in solution is a non-disulfide-linked dimer, the biologically active form of I-309 is monomeric. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine receptor CCR8.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22362
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 6346
Molecular Weight (g/mol) 8.5 kDa
Name Human I-309 (CCL1)
Purification Method Purified
Quantity 2 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias BF534335; C-C motif chemokine 1; C-C motif chemokine ligand 1; CCL 1; Ccl1; CCR8 ligand; chemokine (C-C motif) ligand 1; I-309; inflammatory cytokine I-309; P500; SCYA1; SISe; SIS-epsilon; small inducible cytokine A1; small inducible cytokine A1 (I-309, homologous to mouse Tca-3); small-inducible cytokine A1; T lymphocyte-secreted protein I-309; TCA3; TCA-3; T-cell activation protein 3
Common Name CCL1
Gene Symbol CCL1
Biological Activity Determined by its ability to chemoattract human monocytes population using a concentration range of 10.0-30.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence SKSMQVPFSR CCFSFAEQEI PLRAILCYRN TSSICSNEGL IFKLKRGKEA CALDTVGWVQ RHRKMLRHCP SKRK
Content And Storage -20°C
Activity Determined by its ability to chemoattract human monocytes population using a concentration range of 10.0-30.0 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.