Learn More
Sino Biological Recombinant Human HVEM Protein (ECD, hFc Tag)
Catalog No. 502246928
Encompass_Preferred
A DNA sequence encoding the human TNFRSF14 (NP_003811.2) (Met1-Val202) was expressed with the Fc region of human IgG1 at the C-terminus.
This item is not returnable. View return policy
Catalog No. 50-224-6928 Supplier Sino Biological Supplier No. 10334H02H1
A DNA sequence encoding the human TNFRSF14 (NP_003811.2) (Met1-Val202) was expressed with the Fc region of human IgG1 at the C-terminus.