Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IA-1 Synthetic Peptide

Catalog No. PEP0937 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0937 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0937 Supplier Invitrogen™ Supplier No. PEP0937
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 19 amino acids near the amino terminus of human IA-1. PEP-0937 can be used as a blocking peptide with polyclonal antibody PA5-20823.

IA-1, also known as INSM1, is an essential five zinc-finger transcription factor that while initially identified from an insulinoma cDNA library, is expressed in the developing nervous system. Specifically, IA-1 is expressed as early as E9.5 in mice in the fore-, mid- and hindbrain, spinal cord, retina, and olfactory bulb. It is thought that IA-1 functions as a transcriptional repressor, and is a part of several signaling pathways including those of Notch and sonic hedgehog in addition to that of Ngn3 during pancreatic endocrine cell differentiation. IA-1 is also expressed in multiple tumors, including the majority of neuroendocrine tumors tested so far as well as nearly 100% of all small cell lung carcinomas, indicating that IA-1 may be an important target in cancer therapy.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 3642
Species Human
Purification Method purified
Accession Number Q01101
Common Name INSM1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias IA1; IA-1; INSM transcriptional repressor 1; Insm1; insulinoma associated 1; insulinoma-associated 1; insulinoma-associated protein (IA-1); insulinoma-associated protein 1; insulinoma-associated protein 1-like; LOC100912563; Zinc finger protein IA-1
Gene Symbol INSM1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.