Learn More
BIOHIPPO Human IAP Microsample
SDP
Catalog No. 5000314697
Encompass
Human IAP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IAP in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 106 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IAP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IAP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IAP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14697 Supplier BIOHIPPO Supplier No. BHE15211734
Human IAP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IAP in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 106 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IAP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IAP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IAP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color