Learn More
Sino Biological Human ICAM-2/CD102 Matched ELISA Antibody Pair Set 5 Plates
Supplier: Sino Biological SEK103325
The Human ICAM-2 / CD102 ELISA Pair Set is for the quantitative determination of Human ICAM-2 / CD102. This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for ICAM-2 / CD102 coated on a 96-well plate. Standards and samples are added to the wells, and any ICAM-2 / CD102 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-ICAM-2 / CD102 monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of ICAM-2 / CD102 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.