Learn More
Raybiotech Inc Human ICAM-3 (CD50) ELISA Kit
Supplier:  Raybiotech Inc ELHICAM31
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 65 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human ICAM-3.
Catalog No. 50-194-9366