Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human ICAM-3 Fc Recombinant Protein, PeproTech®

Catalog No. 1502350UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
150-23-50UG 50 μg
15023100UG 100 μg
150-23-10UG 10 μg
150-23-1MG 1 mg
15023250UG 250 μg
15023500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 150-23-50UG Supplier Gibco™ Supplier No. 1502350UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO-derived Recombinant Human ICAM-3 Fc is a glycosylated, disulfide-linked homodimer of 689 amino-acid-residues whose monomer consists of the 456-amino-acid extracellular portion of ICAM-3 fused to the 231-amino-acid length Fc portion of human IgG by two glycine residues. The calculated molecular weight of monomeric CHO cell-derived Recombinant Human ICAM-3 Fc is 75.5 kDa; however, due to glycosylation, it migrates at an apparent molecular weight of approximately 130-140 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CD50 (intracellular adhesion molecule 3, ICAM-3) is a transmembrane glycoprotein expressed by leukocytes, that serves as a counter-receptor for the lymphocyte function-associated antigen (LFA)-1 integrin. Besides functioning as an adhesive molecule that mediates e.g. the contact between T cells and antigen presenting cells, ICAM-3 regulates affinity of LFA-1 for ICAM-1 and induces T cell activation and proliferation. ICAM-3 plays an essential role in the initiation of the immune response both on T cells and antigen presenting cells and interacts also with CD209 (dendritic cell-specific ICAM-3-grabbing nonintegrin, DC-SIGN), a C-type lectin of dendritic cells and macrophages; this process is involved in dialogue between dendritic cells and granulocytes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P32942
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3385
Molecular Weight (g/mol) 75.5 kDa
Name Human ICAM-3 Fc
Purification Method Purified
Quantity 50 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias CD50; CDw50; ICAM3; ICAM-3; ICAM-R; Intercellular adhesion molecule 3; intercellular adhesion molecule-3
Common Name CD50 (ICAM3)
Gene Symbol ICAM3
Biological Activity ICAM-3 Fc supported the adhesion of more than 50% PMA-stimulated HSB2 cells to plates coated with 1.0-4.0 ug/ml ICAM-3 Fc concentration
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence QEFLLRVEPQ NPVLSAGGSL FVNCSTDCPS SEKIALETSL SKELVASGMG WAAFNLSNVT GNSRILCSVY CNGSQITGSS NITVYRLPER VELAPLPPWQ PVGQNFTLRC QVEDGSPRTS LTVVLLRWEE ELSRQPAVEE PAEVTATVLA SRDDHGAPFS CRTELDMQPQ GLGLFVNTSA PRQLRTFVLP VTPPRLVAPR FLEVETSWPV DCTLDGLFPA SEAQVYLALG DQMLNATVMN HGDTLTATAT ATARADQEGA REIVCNVTLG GERREARENL TVFSFLGPIV NLSEPTAHEG STVTVSCMAG ARVQVTLDGV PAAAPGQPAQ LQLNATESDD GRSFFCSATL EVDGEFLHRN SSVQLRVLYG PKIDRATCPQ HLKWKDKTRH VLQCQARGNP YPELRCLKEG SSREVPVGIP FFVNVTHNGT YQCQASSSRG KYTLVVVMDI EAGSSHGGPK SCDKTHTCPP CPAPELLGGP SVFLFPPKPK DTLMISRTPE VTCVVVDVSH EDPEVKFNWY VDGVEVHNAK TKPREEQYNS TYRVVSVLTV LHQDWLNGKE YKCKVSNKAL PAPIEKTISK AKGQPREPQV YTLPPSRDEL TKNQVSLTCL VKGFYPSDIA VEWESNGQPE NNYKTTPPVL DSDGSFFLYS KLTVDKSRWQ QGNVFSCSVM HEALHNHYTQ KSLSLSPGK
Content And Storage -20°C
Activity ICAM-3 Fc supported the adhesion of more than 50% PMA-stimulated HSB2 cells to plates coated with 1.0-4.0 ug/ml ICAM-3 Fc concentration
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.